包裝
pack of 10 ea
製造商/商標名
Cadence Science 6185
一般說明
- Plated brass or 316 stainless steel
- Meets ISO 594 specifications
- Final assembly water tested at 45 PSI
Female Luer Lock (FLL) - contains a tabbed hub for secure fitting to a threaded male Luer lock.
其他客户在看
Global Trade Item Number
货号 | GTIN |
---|---|
CAD6185-10EA | 4061832517537 |