特點
single-channel fixed volume
mechanical
包裝
case of 1 ea
製造商/商標名
BRAND 705862
吸頭容量
(1000 μL)
正在寻找类似产品? 访问 产品对比指南
特點和優勢
- Efficient: one-hand operation
- Flexible: ergonomic design for all hand sizes
- Reliable: volume-change protection prevents accidental adjustments
法律資訊
BRAND is a registered trademark of BRAND GMBH + CO KG
Transferpette is a registered trademark of BRAND GMBH + CO KG
Global Trade Item Number
货号 | GTIN |
---|---|
298069-500ML | |
W236101-10KG | |
W236101-10KG-K | 4061837806605 |
W236101-1KG | |
W236101-5KG-K | 4061837806629 |
W236101-SAMPLE | |
W236128-100G-K | 4061838093264 |
W236128-1KG-K | 4061834403944 |
W236128-25G-K | 4061838191496 |
W236128-SAMPLE | |
74026-100MG | 4061833973370 |
298069-100ML | 4061826656945 |
298069-5ML | |
W236101-1KG-K | 4061837806612 |
W236101-5KG | |
W236101-SAMPLE-K | 4061837509407 |
W236128-100G | |
W236128-1KG | |
W236128-25G | |
W236128-SAMPLE-K | 4061837509414 |
BR705862-1EA | 4061841857105 |