主要文件
BAB608002000
SP Bel-Art® H-B Durac® General Purpose Liquid-In-Glass Thermometer
Red Spirit, temperature -10-200 °C, immersion depth , total
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41112200
NACRES:
NB.79
包裝
pack of 1 ea
製造商/商標名
HB Instruments B60800-2000
參數
-10-200 °C temperature
浸没深度
, total
一般說明
Red Spirit, temperature range-10-200 °C
法律資訊
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Durac is a registered trademark of SP Scienceware, a division of SP
历史批次信息供参考:
分析证书(COA)