V900932
D-(+)-Trehalose dihydrate
Vetec™, reagent grade, ≥99%
Synonym(s):
D-(+)-Trehalose dihydrate, α,α-Trehalose, α-D-Glucopyranosyl-α-D-glucopyranoside
About This Item
Recommended Products
grade
reagent grade
product line
Vetec™
Assay
≥99%
form
powder
storage temp.
room temp
SMILES string
[H]O[H].[H]O[H].OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
InChI
1S/C12H22O11.2H2O/c13-1-3-5(15)7(17)9(19)11(21-3)23-12-10(20)8(18)6(16)4(2-14)22-12;;/h3-20H,1-2H2;2*1H2/t3-,4-,5-,6-,7+,8+,9-,10-,11-,12-;;/m1../s1
InChI key
DPVHGFAJLZWDOC-PVXXTIHASA-N
Looking for similar products? Visit Product Comparison Guide
Application
- Spray-dried microparticles of encapsulated gefitinib for slow-release localized treatment of periodontal disease.: This study explores the development of spray-dried microparticles encapsulating gefitinib using D-(+)-Trehalose dihydrate for slow-release treatment of periodontal disease. The research highlights the potential of this method to provide localized, sustained drug delivery, which can enhance the therapeutic efficacy and reduce systemic side effects (Baldelli et al., 2023).
- On the particle formation of leucine in spray drying of inhalable microparticles.: This research examines the particle formation mechanisms during the spray drying of leucine to create inhalable microparticles. It provides insights into the role ofD-(+)-Trehalose dihydrate in stabilizing the particles and improving their aerosolization properties, which are critical for the development of effective inhalation therapies (Ordoubadi et al., 2021).
Other Notes
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service