Skip to Content
Merck
All Photos(1)

Key Documents

CRM47508

Supelco

Benzoic Acid Solution

certified reference material, 2000 μg/mL in methylene chloride

Synonym(s):

Benzoic acid solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6O2
CAS Number:
Molecular Weight:
122.12
EC Number:
MDL number:
UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Agency

EPA 8270

product line

TraceCERT®

Assay

99.90%

form

liquid

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

2000 μg/mL in methylene chloride

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

density

1.32 g/cm3

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

InChI

1S/C7H6O2/c8-7(9)6-4-2-1-3-5-6/h1-5H,(H,8,9)

InChI key

WPYMKLBDIGXBTP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G C Tremblay et al.
Pharmacology & therapeutics, 60(1), 63-90 (1993-10-01)
Detoxification of sodium benzoate by elimination as a conjugate with glycine, a nonessential amino acid, provides a pathway for the disposal of waste nitrogen. Since 1979, sodium benzoate has been widely used in the therapeutic regimen to combat ammonia toxicity
Benzoic acid intermediates in the anaerobic biodegradation of phenols.
K L Londry et al.
Canadian journal of microbiology, 38(1), 1-11 (1992-01-01)
D J Elder et al.
FEMS microbiology reviews, 13(4), 441-468 (1994-04-01)
Simple homocyclic aromatic compounds are extremely abundant in the environment and are derived largely from lignin. Such compounds may enter anaerobic environments and several groups of bacteria, exhibiting diverse energy-yielding mechanisms, have evolved the capacity to overcome the thermodynamic stability
Production of cis,cis-muconic acid from benzoic acid.
N Yoshikawa et al.
Bioprocess technology, 16, 131-147 (1993-01-01)
Bradley S Moore et al.
Journal of natural products, 65(12), 1956-1962 (2002-12-28)
Although phenylpropanoids and flavonoids are common plant natural products, these major classes of biologically active secondary metabolites are largely absent from bacteria. The ubiquitous plant enzymes phenylalanine ammonia-lyase (PAL) and chalcone synthase (CHS) are key biosynthetic catalysts in phenylpropanoid and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service