Skip to Content
Merck
All Photos(1)

Key Documents

SAB2900392

Sigma-Aldrich

Anti-TANK (AB2) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
clone:
polyclonal
application:
IHC (p)
WB
species reactivity:
mouse, rat, human
technique(s):
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable
citations:
2

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, rat, human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TANK(10010)

Immunogen

Synthetic peptide from human TANK.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hai-ying Chen et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 68(6), 709-713 (2014-08-06)
Exposure of Tca-8113 cells to proteasome inhibitor carbobenzoxy-Leu-Leu-leucinal (MG-132) causing apoptosis is associated with endoplasmic reticulum (ER) stress. X-box-binding protein-1 (XBP1) is an important regulator of a subset of genes active during ER stress, which is related to cell survival
Juhee Jeon et al.
Biochemical pharmacology, 89(4), 515-525 (2014-04-17)
The ligation of interleukin-1 receptor (IL-1R) or tumor necrosis factor receptor 1 (TNFR1) induces the recruitment of adaptor proteins and their concomitant ubiquitination to the proximal receptor signaling complex, respectively. Such are upstream signaling events of IKK that play essential

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service