Skip to Content
Merck
All Photos(3)

Key Documents

527106

Sigma-Aldrich

Nafion perfluorinated resin, aqueous dispersion

greener alternative

10 wt. % in H2O, eq. wt. 1,100

Synonym(s):

D-1021

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
26111700
NACRES:
NA.23

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

10 wt. % in H2O

bp

110 °C

density

1.05 g/mL at 25 °C (lit.)

greener alternative category

InChI

1S/C7HF13O5S.C2F4/c8-1(9)2(10)24-5(15,16)3(11,4(12,13)14)25-6(17,18)7(19,20)26(21,22)23;3-1(4)2(5)6/h(H,21,22,23);

InChI key

FOYUGSIADQEOEK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

As a proton exchange membrane Nafion has been the focus of considerable efforts to develop improved fuel cells
Nafion perfluorinated resin, aqueous dispersion can be used in the surface modification of electrodes for the fabrication of enzyme fuel cells. It can also be used in the formation of a catalyst ink for polymer electrolyte membrane electrolyzer.

Packaging

Packaged in glass bottles

Physical form

Hydrogen ion form

Legal Information

Nafion is a trademark of The Chemours Company FC, LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Switchable aerobic/anaerobic multi-substrate biofuel cell operating on anodic and cathodic enzymatic cascade assemblies
Herkendell K, et al.
Nanoscale, 9(37), 14118-14126 (2017)
State of understanding of nafion.
Kenneth A Mauritz et al.
Chemical reviews, 104(10), 4535-4585 (2005-01-27)
Alternative polymer systems for proton exchange membranes (PEMs).
Michael A Hickner et al.
Chemical reviews, 104(10), 4587-4611 (2005-01-27)
Min-Ho Park et al.
Advanced materials (Deerfield Beach, Fla.), 27(29), 4308-4314 (2015-06-24)
A novel flexible encapsulation method (Flex Lami-capsulation) is reported, which can be applied in the roll-to-roll process for mass production of organic electronic devices. Flex Lami-capsulation is very simple, fast, and getter-free, and is as effective as glass encapsulation. Use

Articles

Materials Issues in Polymer Electrolyte Membrane Fuel Cells

Proton exchange membrane (PEM) fuel cells operate at relatively low temperatures and are composed of two electrodes and a conductive elecrolyte.

Advances in the electrochemical conversion of water to and from hydrogen and oxygen have principally been achieved through the development of new materials and by understanding the mechanisms of the degradation of proton exchange membrane fuel cells (PEMFC) during operation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service