Skip to Content
Merck
All Photos(1)

Key Documents

SAB1408930

Sigma-Aldrich

Monoclonal Anti-DAB2 antibody produced in mouse

clone 3D6, purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3D6, monoclonal

form

buffered aqueous solution

mol wt

antigen 110.22 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DAB2(1601)

General description

DAB2 mRNA is expressed in normal ovarian epithelial cells but is down-regulated or absent from ovarian carcinoma cell lines. The 770-amino acid predicted protein has an overall 83% identity with the mouse p96 protein, a putative mitogen-responsive phosphoprotein; homology is strongest in the amino-terminal end of the protein in a region corresponding to the phosphotyrosine interaction domain. The down-regulation of DAB2 may play an important role in ovarian carcinogenesis. This gene was initially named DOC2 (for Differentially expressed in Ovarian Cancer) and is distinct from the DOC2A and DOC2B genes (for double C2-like domains, alpha and beta). (provided by RefSeq)

Immunogen

DAB2 (AAH03064, 1 a.a. ~ 770 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MSNEVETSATNGQPDQQAAPKAPSKKEKKKGPEKTDEYLLARFKGDGVKYKAKLIGIDDVPDARGDKMSQDSMMKLKGMAAAGRSQGQHKQRIWVNISLSGIKIIDEKTGVIEHEHPVNKISFIARDVTDNRAFGYVCGGEGQHQFFAIKTGQQAEPLVVDLKDLFQVIYNVKKKEEEKKKIEEASKAVENGSEALMILDDQTNKLKSGVDQMDLFGDMSTPPDLNSPTESKDILLVDLNSEIDTNQNSLRENPFLTNGITSCSLPRPTPQASFLPENAFSANLNFFPTPNPDPFRDDPFTQPDQSTPSSFDSLKSPDQKKENSSSSSTPLSNGPLNGDVDYFGQQFDQISNRTGKQEAQAGPWPFSSSQTQPAVRTQNGVSEREQNGFSVKSSPNPFVGSPPKGLSIQNGVKQDLESSVQSSPHDSIAIIPPPQSTKPGRGRRTAKSSANDLLASDIFAPPVSEPSGQASPTGQPTALQPNPLDLFKTSAPAPVGPLVGLGGVTVTLPQAGPWNTASLVFNQSPSMAPGAMMGGQPSGFSQPVIFGTSPAVSGWNQPSPFAASTPPPVPVVWGPSASVAPNAWSTTSPLGNPFQSNIFPAPAVSTQPPSMHSSLLVTPPQPPPRAGPPKDISSDAFTALDPLGDKEIKDVKEMFKDFQLWQPPAVPARKGEQTSSGTLSAFASYFNSKVGIPQENADHDDFDANQLLNKINEPPKPAPRQVSLPVTKSTDNAFENPFFKDSFGSSQASVASSQPVSSEMYRDPFGNPFA

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service