Skip to Content
Merck
All Photos(1)

Key Documents

MABC1176M

Sigma-Aldrich

Anti-Bax Antibody, clone 6A7

clone 6A7, from mouse

Synonym(s):

Apoptosis regulator BAX, Bcl-2-like protein 4, Bcl2-L-4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6A7, monoclonal

species reactivity

human

species reactivity (predicted by homology)

rat (based on 100% sequence homology), mouse (based on 100% sequence homology)

technique(s)

immunofluorescence: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... BAX(581)

General description

Apoptosis regulator Bax (UniProt: Q07812; also known as Bcl-2-like protein 4, Bcl2-L-4) is encoded by the BAX (also known as BCL2L4) gene (Gene ID: 581) in human. Bax is a pro-apoptotic member of the Bcl-2 family of proteins. It displays extensive homology with Bcl-2 and is variably expressed in different cells, usually at very low concentrations in normal cells. Upon induction of apoptosis Bax undergoes a conformational change and translocates from cytosol to mitochondrial membrane. Bax forms homodimers as well as heterodimers with Bcl-2 through common binding motif and the ratio of Bax to Bcl-2 determines the survival or death of cells during apoptosis. BAX is reported to interact with, and increase the opening of, the mitochondrial voltage-dependent anion channel (VDAC), which leads to the loss in membrane potential and the release of cytochrome c. Multiple alternatively spliced transcript variants, which encode different isoforms, have been reported for this gene. Clone 6A7 reacts with multiple isoforms of Bax.

Specificity

Clone 6A7 recognizes BAX in human cells. It targets an epitope with in 13 amino acids from the N-terminal region.

Immunogen

KLH-cojugated linear peptide coprresponding to 13 amino acids from the N-terminal region of human BAX.

Application

Anti-Bax, clone 6A7, Cat. No. MABC1176, is a mouse monoclonal antibody that detects Bax and has been tested for use in Immunofluorescence, Immunoprecipitation, and Western Blotting.
Research Category
Apoptosis & Cancer
Western Blotting Analysis: A representative lot detected Bax in Western Blotting applications (Xin, M., et. al. (2014). Nat Commun. 5:4935).

Immunofluorescence Analysis: A representative lot detected Bax in Immunofluorescence applications (Xin, M., et. al. (2014). Nat Commun. 5:4935).

Immunoprecipitation Analysis:A representative lot detected Bax in Immunoprecipitation applications (Xin, M., et. al. (2014). Nat Commun. 5:4935; Hsu, Y.T., et. al. (1998). J Biol Chem. 273(17):10777-83).

Quality

Evaluated by Immunoprecipitation in THP-1 cell lysates.

Immunoprecipitation Analysis: A 1:250 dilution of this antibody detected Bax in THP-1 cell lysates.

Target description

~15 kDa observed; 21.18 kDa calculated. Uncharacterized bands may be observed in some lysate(s). Multiple isoforms for this target (8), ranging in MWs between 4-24 kDa, are reported. Most of the predicted MWs reside between 12-19 kDa Based on the sequence it is expected to react with isoforms 1,2,4,5, and 8.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service