Skip to Content
Merck
All Photos(1)

Key Documents

SRP6063

Sigma-Aldrich

PIN1 human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE)

Synonym(s):

DOD, NIMA-interacting protein 1, PPIase Pin1, Rotamase Pin1, UBL5

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥95% (SDS-PAGE)

form

liquid

mol wt

18.2 kDa (163 aa)

packaging

pkg of 100 μg

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PIN1(5300)

General description

PIN1 (peptidylprolyl cis/trans isomerase, NIMA-interacting 1) is a peptidyl prolyl cis-trans isomerase. It is a conserved eukaryotic protein that contains an amino-terminal WW domain, which functions as a phosphorylated serine or threonine residue-binding site. Its C-terminal domain functions as the enzymatic domain, and is responsible for catalyzing the cis/trans isomerization of pSer/Thr-Pro bonds.

Biochem/physiol Actions

PIN1 (peptidylprolyl cis/trans isomerase, NIMA (never in mitosis A)-interacting 1) associates specifically with the phosphoserine-proline or phosphothreonine-proline residues immediately preceding proline (pSer/Thr-Pro), and promotes cis/trans isomerization of the peptide bond. This protein is involved in the amplification of the phosphorylation signaling, and is involved in catalyzing target dephosphorylation, control of protein stability and ubiquitination. It also mediates the cellular localization of its target proteins. It is highly up-regulated in multiple types of cancers, such as breast and prostate cancer, and is essential for the functionality and cross-talk of tumorigenic pathways. It is also implicated in Alzheimer′s disease (AD) and asthma.

Physical form

1 mg/mL solution in 20 mM Tris-HCl buffer (pH 7.5) containing 100 mM NaCl, 5 mM DTT, 20% glycerol.

Preparation Note

Centrifuge the vial prior to opening.

Other Notes

MADEEKLPPG WEKRMSRSSG RVYYFNHITN ASQWERPSGN SSSGGKNGQG EPARVRCSHL LVKHSQSRRP SSWRQEKITR TKEEALELIN GYIQKIKSGE EDFESLASQF SDCSSAKARG DLGAFSRGQM QKPFEDASFA LRTGEMSGPV FTDSGIHIIL RTE

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guo-Liang Huang et al.
Cell death & disease, 7(12), e2571-e2571 (2016-12-30)
The function of activating transcription factor 1 (ATF1) and the mechanism about why ATF1 was over-phosphorylated in nasopharyngeal carcinoma (NPC) progression is completely undiscovered. In this study, a series of experiments both in vitro and in vivo were used to
Pin1 is related with clinical stage of papillary thyroid carcinoma.
Jiang L et al
World Journal of Surgical Oncology, 14, 95-95 (2016)
Peptidyl-Prolyl cis/trans Isomerase NIMA-Interacting 1 as a Therapeutic Target in Hepatocellular Carcinoma.
Kim G et al
Biological & Pharmaceutical Bulletin, 38(7), 975-979 (2015)
Jin Won Yang et al.
Journal of hepatology, 60(6), 1235-1241 (2014-02-18)
Therapeutic management of liver fibrosis remains an unsolved clinical problem. Hepatic accumulation of extracellular matrix, mainly collagen, is mediated by the production of transforming growth factor-β1 (TGF-β1) in stellate cells. Pin1, a peptidyl-prolyl isomerase, plays an important pathophysiological role in
Garam Kim et al.
Molecular carcinogenesis, 54(6), 440-448 (2013-11-23)
Pin1, a conserved eukaryotic Peptidyl-prolyl cis/trans isomerase, has profound effects on numerous key-signaling molecules, and its deregulation contributes to disease, particularly cancer. Although Pin1-mediated prolyl isomerization is an essential and novel regulatory mechanism for protein phosphorylation, little is known about

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service