Merck
EP5490035003
Rotor for Eppendorf® Concentrator Plus
F-45-36-15, non aerosol tight, to hold, 36 x 3.0/5.0 mL flat-bottom tubes
About This Item
UNSPSC-Code:
41103909
NACRES:
NB.82
Leistungsmerkmale
to hold 36 x 15 x 48 mm
to hold 36 x 3.0/5.0 mL flat-bottom tubes
fixed angle rotor
non aerosol tight
Verpackung
pack of 1 ea
Hersteller/Markenname
Eppendorf® 5490035003 (EU)
Methode(n)
centrifugation: suitable
Kompatibilität
for use with centrifuge models Concentrator Plus
Allgemeine Beschreibung
Selection of rotors for the Eppendorf Concentrator Plus.
Rechtliche Hinweise
Eppendorf is a registered trademark of Eppendorf AG