Merck
Z568988
Sigma-Aldrich
Aldrich® Liebig condenser with cup
Overall H 200 mm, joint: ST/NS 24/40
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
Verbindung
joint: ST/NS24/40
Gesamt H
200 mm
Verwandte Kategorien
Allgemeine Beschreibung
Condensate cup catches water that forms on the outer surface of the condenser during use and eliminates puddling on reactor and equipment. Tubing connects to cup′s hose barb to drain away water automatically.
Rechtliche Hinweise
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Global Trade Item Number
SKUGTIN
Z568988-1EA4061832119908