Merck
Z531758
Sigma-Aldrich
Tubing Adapters with Torion Connection
With hose barb, O.D. 5/16 in., male joint: ST/NS 14/20
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.35
Verbindung
male joint: ST/NS14/20
AD
5/16 in.
7.9 mm
Allgemeine Beschreibung
The unique Torion connection ensures an airtight seal between tubing and reaction vessels when transferring air-sensitive materials.
5/16 in. O.D fits a #8 Torion, 1/2 in. O.D. fits a #14 Torion, and 3/16 in. O.D fits a #5 Torion.
  • Hard-wall transfer tubing (e.g. PP or PTFE) is inserted through the threaded polypropylene bushing and PTFE-coated sealing ring of the adapter
  • Turn the bushing to make an airtight compression seal with the tubing
  • Use the optional solid-glass plug to make an airtight Torion seal when the transfer tubing is not in use
  • O.D. refers to the tubing outer diameter
Rechtliche Hinweise
Torion is a trademark of Bibby Sterlin Ltd.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)