Merck
Z189022
Sigma-Aldrich
Ace four-neck round-bottom flask
capacity 2,000 mL
Synonym(e):
bottom flask, flask round, flask round bottom
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.43
Materialien
borosilicate glass
four-neck
round bottom flask
Verbindung
90 degree angle joint ST/NS24/40
center joint ST/NS34/45 vertical
side Joints ST/NS24/40 vertical, except for 500 mL, which is angled (2)
Sterilität
non-sterile
Leistungsmerkmale
closure type Standard Taper Joint
Verpackung
pack of 1
Hersteller/Markenname
Ace Glass 6952-28
Kapazität
2,000 mL
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)