Merck
SYNS511740
Synthware T-bore glass stopcock Entire assembly
bore size 4 mm, plug size 17/40
Synonym(e):
Synthware S511740, T-shape stopcock
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.51
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware S511740
Bohrungsgröße
4 mm
Allgemeine Beschreibung
With solid glass plug. With constant pressure retention spring and clip.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)