Merck
SYNA781410
Synthware thermometer adapter with hose connection
joint: ST/NS 14/10
Synonym(e):
Synthware A781410, flow control adapter
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.51
Verbindung
joint: ST/NS14/10
Verpackung
pack of 1 ea
Hersteller/Markenname
A781410
Verwandte Kategorien
Allgemeine Beschreibung
Synthware inlet or thermometer adapter with a compression cap and a Viton® O-ring to form a tight seal. Cap accommodates a 5.0-7.5mm O.D. plain stem thermometer. Supplied with one Nylon cap and one Viton® O-ring. To use as syringe port, replace O-ring with PTFE-lined septum.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)