Merck
DWK212174405
DURAN® Super duty Erlenmeyer narrow-neck flasks
flask capacity (500 mL)
Synonym(e):
212174405
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.17
Materialien
flat
Kompatibilität mit Sterilisationsverfahren
autoclavable compatible
Leistungsmerkmale
without cap
Verpackung
case of 10 ea
Hersteller/Markenname
DURAN® 212174405
Kolbenkapazität
(500 mL)
Allgemeine Beschreibung
Higher impact strength, mechanical strength and stability achieved by reinforced rim. Economical due to longer life span and outstanding corrosion protection. White easy-to-read scale with graduation marks to estimate the content. Fired-on, highly durable white ceramic print.
Sonstige Hinweise
Rechtliche Hinweise
DURAN is a registered trademark of DWK Life Sciences
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)