BR150915
BRAND® watch glass
PTFE, diam. 100 mm
Synonym(e):
Dishes, PTFE, Watch glasses
Alle Fotos(2)
About This Item
UNSPSC-Code:
41121801
NACRES:
NB.43
Empfohlene Produkte
Materialien
PTFE
Hersteller/Markenname
BRAND 150915
Durchmesser
100 mm
Verwandte Kategorien
Allgemeine Beschreibung
The BRAND® watch glass is made from high-quality PTFE, making it ideal for covering PTFE beakers. Its excellent chemical resistance ensures durability and reliability when handling a wide range of substances. This watch glass provides a secure cover to prevent contamination and evaporation during experiments. Its lightweight and non-reactive properties make it a valuable accessory in laboratory settings, enhancing the efficiency of various applications. This versatile tool is essential for maintaining the integrity of samples and solutions in the lab.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
BR150915-1EA | 4061831854107 |