Merck
BR140006
BRAND® separating funnel with screw cap
size 500 mL
Synonym(e):
Funnels, Separating funnels
About This Item
UNSPSC-Code:
41121809
NACRES:
NB.43
Materialien
conical PTFE stopcock
polypropylene
Verpackung
pack of 1 ea
Hersteller/Markenname
BRAND 140006
Größe
500 mL
Allgemeine Beschreibung
Polypropylene, high clarity for observation of contents. With polypropylene screw cap and special conical PTFE stopcock with retention device. Autoclavable (121 °C).
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)