BR13903
BRAND® compact SILBERBRAND burette, with PTFE stopcock
volume 10 mL
Synonym(e):
Burettes, Compact burettes, SILBERBRAND
Alle Fotos(3)
About This Item
UNSPSC-Code:
41121808
NACRES:
NB.43
Empfohlene Produkte
Materialien
glass
Hersteller/Markenname
BRAND 13903
Messbereich Volumen
10 mL, accuracy: ±0.05 mL
Volumen
10 mL
Verwandte Kategorien
Allgemeine Beschreibung
The New Generation of Burettes from BRAND. With PTFE stopcock, fast to dismantle and easy to clean. Simple repair - all individual components replaceable. Boro 3.3, removable PTFE stopcock. Error limits according to class B, DIN EN ISO 385. Calibrated to deliver (TD, Ex).
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
BR13903-1EA | 4061831853131 |