Merck
BAF131641923
Scienceware® biohazard disposal bags
width × H × thickness 480 mm × 23 in. (580 mm) × 0.04 mm (1.5 mil), pack of 200 ea, red
Synonym(e):
Scienceware® biohazard disposal bags
About This Item
UNSPSC-Code:
41122800
NACRES:
NB.35
Materialien
red
Verpackung
pack of 200 ea
Hersteller/Markenname
Bel-Art F13164-1923
Kapazität
18.9-34.1 L (5-9 g)
width × H × thickness
480 mm × 23 in. (580 mm) × 0.04 mm (1.5 mil)
Allgemeine Beschreibung
Bright red polypropylene bags include black biohazard symbol and precautionary procedures printed in four languages (English, Spanish, French, German) for added safety.
Leistungsmerkmale und Vorteile
  • Sterilization indicator patch darkens when exposed to steam sterilization for 20 minutes
  • High temperature steam autoclavable to 135°C (275°F)
  • Pass ASTM 1922 Tear Resistance and ASTM 1709 Dart Impact Tests
  • Made in USA
Rechtliche Hinweise
Scienceware is a registered trademark of SP Scienceware, a division of SP
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)