Skip to Content
Merck
All Photos(6)

Key Documents

CWA-1084

Sigma-Aldrich

Anti-VCAM1 antibody produced in mouse

greener alternative

clone 1.G11B1, ColorWheel Dye-Ready

Synonym(s):

Vascular cell adhesion protein 1;V-CAM 1;VCAM-1;INCAM-100

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

Product Name

Anti-Human CD106 (1.G11B1) ColorWheel® Dye-Ready mAb,

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

1.G11B1, monoclonal

product line

ColorWheel®

form

lyophilized

mol wt

calculated mol wt 81.28 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

porcine

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

flow cytometry: suitable

isotype

IgG1κ

epitope sequence

Extracellular domain

Protein ID accession no.

UniProt accession no.

compatibility

for use with ColorWheel® Dyes (Required, (Sold Separately))

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

mouse ... VCAM1(7412)

General description

ColorWheel® technology is a novel and proprietary method of creating your own antibody and dye combinations for use in flow cytometry. By incubating any ColorWheel® antibody with any ColorWheel® dye, researchers can quickly and simply produce primary conjugated antibodies for use in single color or multicolor flow cytometry analysis. Each ColorWheel® antibody and dye is lyophilized for long-term storage, allowing for simplicity and flexibility without compromise. ColorWheel® technology requires both ColorWheel® antibody and ColorWheel® dye for Flow Cytometry application.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This product is Preservative-free, lyophilized product for enhanced stability and allow for ambient shipping and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.

Specificity

Clone 1.G11B1 is a mouse monoclonal antibody that detects CD106. It targets an epitope within the extracellular domain.

Immunogen

Human endothelial cells

Application

Quality Control Testing

Evaluated by Flow Cytometry in HUVEC .

Flow Cytometry Analysis: Staining of HUVEC was performed using 5 μL of a 1:1 mixture of Cat. No. CWA-1084, Anti-Human CD106 (1.G11B1) ColorWheel® Dye-Ready mAb and Cat. No. CWD-PE ColorWheel® Antibody-Ready Phycoerythrin (PE) Dye or an equivalent amount of PE-conjugated Mouse IgG1 isotype control.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Vascular cell adhesion protein 1 (UniProt: P19320; also known as V-CAM 1, VCAM-1, INCAM-100, CD106) is encoded by the VCAM1 gene (Gene ID: 7412) in human. CD106 is a single-pass type I membrane glycoprotein that is synthesized with a signal peptide (aa 1-24), which is subsequently cleaved off to generate the mature form that contains an extracellular domain (aa 25-698), a transmembrane domain (aa 699-720), and a cytoplasmic domain (aa 721-739). The extracellular domain contains seven Ig-like C2-type domains. Ig-like domain pairs of 1 and 4, 2 and 5, and 3 and 6 are highly homologous with each other. It is predominantly expressed in endothelial cells but is reported to be absent on resting endothelial cells. Endothelial cells respond to IL-1 and TNF-a by upregulating the expression of CD106, with maximal activity reaching at 6 to 12 hours following exposure. Under high degree of inflammation and chronic conditions in some diseases, CD106 expression is also observed on the surface of other cells, including tissue macrophages, dendritic cells, bone marrow fibroblasts, myoblasts, oocytes, Kupffer cells, Sertoli cells, and cancer cells. CD106 functions in leukocyte-endothelial cell adhesion. It interacts with integrin a4/b1 on leukocytes and mediates both adhesion and signal transduction. It supports leukocyte rolling that allows exposure to chemokines and other chemotactic signaling molecules that are secreted by the endothelium or adjacent cells. These events can also trigger intracellular signaling and functional activation of leukocyte integrins through G protein-linked receptors. (Ref.: Kong, DH., et al. (2018). Int. J. Mol. Sci. 19(4); 1057; Carter, RA., and Wicks, IP. (2001). Arthritis Rheum. 44(5); 985-994).

Physical form

Lyophilized from PBS containing D-Mannitol and Sucrose, normal appearance is a dried pellet. Reconstituted antibody solution is stable and functional as assessed by functional testing. Contains no biocide or preservatives, such as azide.

Reconstitution

1.0 mg/mL after reconstitution at 25 μL in PBS. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C. Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial. Reconstitute each vial by adding 25 μL PBS. Protect from light.

Legal Information

ColorWheel is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

required but not provided

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service