Skip to Content
Merck
All Photos(1)

Key Documents

757748

Sigma-Aldrich

Poly(ethylene glycol) [N-(2-maleimidoethyl)carbamoyl]methyl ether 2-(biotinylamino)ethane

average Mn 5,400

Synonym(s):

Biotin polyoxyethylene maleimide, Biotin-PEG, Biotin-PEG-maleimide, Biotinylated PEG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H16N3O2S(C2H4O)nC8H9N2O3
UNSPSC Code:
12162002
NACRES:
NA.23

form

solid

mol wt

PEG average Mn 5,000 (n~110)
average Mn 5,400

reaction suitability

reagent type: cross-linking reagent
reactivity: thiol reactive

mp

47-52 °C

Ω-end

maleimide

α-end

biotin

polymer architecture

shape: linear
functionality: heterobifunctional

storage temp.

−20°C

Application

Applications include: bioconjugation, drug delivery, PEG hydrogel, and crosslinker.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kwang-Hyun Park et al.
Nature communications, 11(1), 2728-2728 (2020-06-03)
The Pseudomonas putida phenol-responsive regulator DmpR is a bacterial enhancer binding protein (bEBP) from the AAA+ ATPase family. Even though it was discovered more than two decades ago and has been widely used for aromatic hydrocarbon sensing, the activation mechanism

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service