Skip to Content
Merck
All Photos(1)

Key Documents

421790

Sigma-Aldrich

(1R,2R)-trans-1,2-Cyclohexanediol

99%

Synonym(s):

(1R)-trans-1,2-Cyclohexanediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H10(OH)2
CAS Number:
Molecular Weight:
116.16
Beilstein:
4780913
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

optical activity

[α]20/D −39°, c = 1.6 in H2O

optical purity

ee: 99% (HPLC)

mp

107-109 °C (lit.)

SMILES string

O[C@@H]1CCCC[C@H]1O

InChI

1S/C6H12O2/c7-5-3-1-2-4-6(5)8/h5-8H,1-4H2/t5-,6-/m1/s1

InChI key

PFURGBBHAOXLIO-PHDIDXHHSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elena Parmigiani et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 35(19), 7388-7402 (2015-05-15)
Cerebellar GABAergic interneurons in mouse comprise multiple subsets of morphologically and neurochemically distinct phenotypes located at strategic nodes of cerebellar local circuits. These cells are produced by common progenitors deriving from the ventricular epithelium during embryogenesis and from the prospective

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service