SU861027
Super Clean (Base-Plate Design) Gas Purifier
triple trap (hydrocarbon, moisture, oxygen), for helium carrier gas
About This Item
Codice UNSPSC:
41115717
NACRES:
SB.54
Descrizione generale
Gas purifier helps in enhancing the performance of GC detectors. It can reduce the background signal in flame ionization and nitrogen phosphorus detectors.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Gas Chromatographic Techniques and Applications
Alan John Handley, E. R. Adlard
Science, 135-135 (2001)
Global Trade Item Number
SKU | GTIN |
---|---|
SU861027 | 4061837328794 |
SU861040 | 4061837328800 |