Documenti fondamentali
58539-U
Supelco
SSI TC-20 Tubing Cutter
AC/DC input 110 V / 220 V AC, 50-60 Hz (voltage selectable), CE compliant
Sinonimo/i:
Tubing Cutter Tool
About This Item
Codice UNSPSC:
41115700
NACRES:
SB.52
Caratteristiche
CE compliant
Descrizione generale
The SSI Model TC-20 electric stainless steel tube cutter assures a zero dead volume connection. Because the tubing is held securely in a clamp vise on the vertical swing arm, a square cut is produced when the swing arm is lowered against the abrasive cutting wheel, which produces a finished end. No lubricant or cutting fluid is required. The unit, which is CE marked, will cut most common tubing used in chromatography. Tubing with 1/16 in., 1/8 in., and 1/4 in. outside diameter, can be smooth-cut and dressed without distortion.
The precision ground dressing tool for the 1/16 in. O.D. tubing is included and is attached directly to the swing arm: it cannot be misplaced or lost. A dressing tool (deburring tool - Cat. No. 58804) for 1/8 in. diameter can be ordered separately.
Prodotti consigliati
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Note legali
SSI is a trademark of Scientific Systems, Inc.
Raccomandato
N° Catalogo
Descrizione
Determinazione del prezzo
Ricambio
N° Catalogo
Descrizione
Determinazione del prezzo
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 2
1 of 2
Global Trade Item Number
SKUGTIN
58539-U4061833518625