Documenti fondamentali
58468
Supelco
Dynaseal Fittings
pkg of 10 ea, Kel-F ferrule
Sinonimo/i:
Kel-F FERRULES,1/16" PK/10
About This Item
Codice UNSPSC:
41115700
Materiali
Kel-F ferrule
Confezionamento
pkg of 10 ea
Raccordi
1/16 in. Dynaseal ferrule for 1/16 in. O.D. tubing
Caratteristiche e vantaggi
Dynaseal hand-tight fittings will seal against pressures to over 7000psi/490kg/cm². Each fitting consists of a reusable nut, collet, and ferrule. The polymer ferrule will not deform stainless steel seats and can be reused. Except for the fitting for Rheodyne valves, these fittings are compatible with all 1/16 in. female compression-type fittings now used on HPLC columns.
Prodotti consigliati
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Note legali
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)