Documenti fondamentali
56030
Omnifit® Standard Bore Connector
Two-way connectors
About This Item
Codice UNSPSC:
12000000
tecniche
LPLC: suitable
Foro
1.5 mm
Compatibilità
for use with 1/16 in. tubing
Descrizione generale
Omnifit® Standard Bore Connector is very versatile in nature and is used for connecting various types of hardwall tubings. The standard bore connector has a manual valve operation on each end, thus the flow can be switched on or off as required. It is primarily used for fluid-line interconnection, sampling and mixing.
Applicazioni
Omnifit® Standard Bore Connector was used to construct the flow-injection system for amperometric determination of tacrine.
Caratteristiche e vantaggi
O-rings and PTFE cone included.
Note legali
Omnifit is a registered trademark of Diba Industries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Flow-Injection Amperometric Determination of Tacrine based on Ion Transfer across a Water-Plasticized Polymeric Membrane Interface.
Joaquin, A. C.
Sensors, 7, 1185-1192 (2007)
Modern Instrumental Techniques
Keith, A
Soil and Environmental Analysis, 1-700 (2003)