Key Documents
41909
Supelco
Stand for SCOTTY® 110 HP Cylinder
About This Item
Codice UNSPSC:
41116105
Confezionamento
pkg of 1 ea
applicazioni
environmental
Descrizione generale
Ensures your SCOTTY 110 cylinder will be stable on a bench-top or other flat surface. This product will hold cylinders with a 3¼" diameter.
Applicazioni
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Note legali
SCOTTY is a registered trademark of Air Liquide
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)