Documenti fondamentali
22375
Chart Paper for Recorder
Blank for Spectra Physics 4270, 4290 and Varian Vista 401/402, box of 25
About This Item
Codice UNSPSC:
14111500
Confezionamento
box of 25
Largh. × lungh.
9.3 in. × 170 ft
Applicazioni
Analog signal from the chromatograph may be connected to a recorder with chart paper speed of 5 min/in. during an analysis to measure Glutethimide concentrations in the serum and cerebrospinal fluid (CSF) using gas chromatography.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
John Kenkel
Analytical Chemistry for Technicians, 366-366 (1994)
A.V.Bakshi U.A.Bakshi
Electronic Measurements & Instrumentation (2008)
Comparison of glutethimide concentration in the serum and cerebrospinal fluid of humans in drug overdose.
M Gold et al.
Clinical chemistry, 19(10), 1158-1161 (1973-10-01)