Documenti fondamentali
22054
Swagelok® PTFE Ferrules
PTFE front ferrule, Swagelok for 1/4 in. tubing
About This Item
Codice UNSPSC:
41115717
Materiali
PTFE front ferrule
Confezionamento
pkg of 10 ea
Diam. int.
1/4 in.
Raccordi
Swagelok for 1/4 in. tubing
Descrizione generale
Swage type fittings are one of the most popular connectors which are available in metal and plastic forms; they are very useful for connecting similar or dissimilar materials, like metal to glass or metal to plastic. The general swage tubing coupler has four parts namely the body, into which the tube is placed, two ferrules that is front ferrule and back ferrule and lastly a packing nut.
Use PTFE ferrules instead of brass or stainless steel ferrules when attaching fittings to glass tubing, or to flexible tubing (such as PTFE or Tygon).
Applicazioni
Swagelok® PTFE ferrules maybe used, for making all the connections between glass and PTFE present in the containment vessels, piping, and sampling equipment in a bench scale biofilter system, in an experimental study investigating the effects of periods of non-use on biofilter performance.
Note legali
Swagelok is a registered trademark of Swagelok Company
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 4
1 of 4
Swagelok® Tee Swagelok® 400-3, stainless steel, 1/4 in. Swagelok
22036
Swagelok® Tee
PTFE Tubing tubing L × O.D. × I.D. 25 ft × 1/4 in. (6.35 mm) × 0.228 in. (5.8 mm)
Supelco
20533
PTFE Tubing
Effect of periods of non-use on biofilter performance.
Jason MF and Loehr RC.
Journal of the Air & Waste Management Association (1995), 46 (6), 539-546 (1996)
The Manipulation of Air-Sensitive Compounds.
Shriver DF.
Nature, 217-218 (1986)