Documenti fondamentali
Z757403
Sartorius Proline® Plus mechanical pippettor
single channel, volume 100-1000 μL
Sinonimo/i:
Mechanical Pipettor, Proline Plus Pipettor
About This Item
Codice UNSPSC:
41121500
NACRES:
NB.01
Caratteristiche
electronic: no
single-channel
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Sartorius 728070
Volume
100-1000 μL
Descrizione generale
The Sartorius Proline Plus pipettes of the Biohitfamily are high quality with a light pipetting force. With robust design, the Proline Plus is suitable for variable every day laboratory applications and covers the full range of 0.1μL to 10 mL.
  • Reduced pipetting forces prevent RSI and improve the results
  • Accurate and precise
  • Contamination free pipetting with Safe-Cone Filters
  • Robust design
  • Easiest pipette to maintain
Note legali
Proline is a registered trademark of Sartorius Biohit Liquid Handling Oy
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)