Z710962
Rack for glass slides
for 20/40 slides, glass
Sinonimo/i:
900304
About This Item
Codice UNSPSC:
41122800
NACRES:
MA.01
Prodotti consigliati
Materiali
glass
Caratteristiche
Holds 20/40 x slides 76x26 mm
Produttore/marchio commerciale
WHEATON® 900304
Misura
118x88x70 mm
Descrizione generale
Glass rack holds 20 single slides, 39 slides arranged alternately straight across and diagonally, or 40 slides back to back. Manufactured from soda-lime glass.
Note legali
WHEATON is a registered trademark of Wheaton Industries, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)