Documenti fondamentali
Z709093
ART® non-filtered tip
HLT 1000 SoftFit, size 100-1000 μL, Pure packed, sterile
Sinonimo/i:
pipet tips, pipette tips
About This Item
Codice UNSPSC:
41121600
eCl@ss:
32079201
NACRES:
NB.01
Volume massimo:
1 mL
Misura:
100-1000 μL
Sterilità:
sterile
Caratteristiche:
DNase free
RNase free
Pure packed
Confezionamento:
pack of 800 ea
Produttore/marchio commerciale:
Molecular BioProducts 3581
Sterilità
sterile
Caratteristiche
DNase free
RNase free
Pure packed
Confezionamento
pack of 800 ea
Produttore/marchio commerciale
Molecular BioProducts 3581
Volume massimo
1 mL
Misura
100-1000 μL
Descrizione generale
SoftFit design makes loading and ejecting easy and provides a universal fit.
Molecular BioProducts Pure lines of pipette tips are pre-sterilized and packaged in a controlled environment to eliminate contamination. These tips are tested by an independent lab to ensure that they meet industry standards. It is ideal for a wide variety of applications.
Note legali
ART is a registered trademark of Molecular BioProducts, Inc.
SoftFit-L is a trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)