Documenti fondamentali
Z659312
Bandelin SONOPULS ultrasonic homogenizer accessories
Sleeve adapter NA 29G for ground glass vessels
Sinonimo/i:
ultrasonic homogenizer adapter
About This Item
Codice UNSPSC:
41101520
NACRES:
NB.74
Materiali
PTFE
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Bandelin 540
Disponibilità
available only in EU
Descrizione generale
The standard ground joint adapter is used to create a tight connection between laboratory vessels with standard ground joints and SONOPULS ultrasonic homogenizers. This enables a wide range of sound irradiation setups and flow-through ultrasonic reactors to be realized. The adapter is screwed onto the external thread of the SONOPULS standard or booster horn.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)