Documenti fondamentali
Z382077
Sigma-Aldrich
Nalgene® Lowboy carboy with spigot
nominal capacity 8 L (2 gal), polypropylene copolymer bottle (PPCO)
Sinonimo/i:
carboys
About This Item
Codice UNSPSC:
41121820
NACRES:
NB.35
Materiali
polypropylene copolymer bottle (PPCO)
polypropylene spigot and screw closure
Descrizione
Not recommended for use with most organic solvents
Produttore/marchio commerciale
Nalgene® 2324 style
Lungh. × Largh. × Alt.
6 1/2 in. (165 mm) × 14 1/2 in. (368 mm) × 12 in. (305 mm)
Capacità nominale
8 L (2 gal)
Descrizione generale
Fits into tight places and can be stacked three high. Integral handgrip for safe and easy transport. Tolerant of strong acids and bases.
Similar to Style 2323, but all polypropylene.
Resistenza chim/fis
Autoclavable
Note legali
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)