Documenti fondamentali
Z360295
Carboy spigots
Tefzel®
Sinonimo/i:
plastic spigot, plastic spigots
About This Item
Codice UNSPSC:
41121820
NACRES:
NB.35
Materiali
Tefzel®
Produttore/marchio commerciale
Nalgene®
Descrizione generale
Spigots fit all Nalgene carboys. Both are autoclavable when installed on autoclavable carboys. Thread is 11/8 in. - 12 straight female threads. Spout accepts 5/8 in. I.D. tubing.
Tefzel® ETPE for use with solvents and other chemicals where a PP spigot is unsatisfactory.
Note legali
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Tefzel is a registered trademark of E. I. du Pont de Nemours and Company
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)