Z358290
Nalgene® staining box
L × W × H 12.5 cm × 12.5 cm × 5 cm
Sinonimo/i:
electrophoresis tray, gel staining box
About This Item
Codice UNSPSC:
41123300
NACRES:
NB.22
Prodotti consigliati
Materiali
(PCTG)
Confezionamento
pack of 2 ea
Produttore/marchio commerciale
Nalgene® 5705-1010
Lungh. × Largh. × Alt.
12.5 cm × 12.5 cm × 5 cm
Descrizione generale
Constructed of PCTG [poly(1,4-cyclohexylene dimethylene terephthalate)glycol] with a polypropylene copolymer cover and PDPE plug. Designed for staining and destaining electrophoresis gels. Resists temperatures from −70 to +70°C and is resistant to acids, bases, and some organic solvents. Plug allows draining without handling gels. Stackable.
Note legali
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)