WPB01341WA
Whirl-Pak® Flat Wire, Write-On Bag
532 mL capacity(18 oz)
Sinonimo/i:
sample bag with label, sampling bag
About This Item
Codice UNSPSC:
24111500
NACRES:
NB.35
Prodotti consigliati
Confezionamento
pkg of 500 ea
Produttore/marchio commerciale
B01341WA (Nasco)
Largh. × lungh. × spess.
11.5 cm × 23 cm × 0.064 mm
Capacità
532 mL × 18 oz
532 mL capacity(18 oz)
Categorie correlate
Descrizione generale
These bags are made with one flat wire, rather than the traditional round wires. The flat wire provides a thicker tab, resulting in a heavier closure which is preferred in some applications. The aluminum wire features “PUNCTURE PROOF TABS,” so the wire ends are completely covered, eliminating any sharp, cutting, exposed edges. The aluminum will not rust when coming in contact with water or other liquids.
The white, write-on strip makes sample identification easier and more convenient. It′s easy to write on this white strip using an ordinary ballpoint pen, pencil, or felt-tip marker. The paper-like finish virtually eliminates ink smearing, and provides a surface that will accept a variety of writing instruments.
The white, write-on strip makes sample identification easier and more convenient. It′s easy to write on this white strip using an ordinary ballpoint pen, pencil, or felt-tip marker. The paper-like finish virtually eliminates ink smearing, and provides a surface that will accept a variety of writing instruments.
Note legali
Whirl-pak is a registered trademark of The Aristotle Corporation
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Global Trade Item Number
SKU | GTIN |
---|---|
WPB01341WA-500EA | 4061833567760 |