WHA77002803
Whatman® UNIFILTER® 96 well filter plates
clear polystyrene, GF/B membrane, non-sterile, biomolecule, 25 ea
Sinonimo/i:
Z382337, micro plate, multiwell plate, well plates
About This Item
Prodotti consigliati
Nome del prodotto
Whatman® UNIFILTER® plates, working volume 800 μL, with Whatman GF/B membrane
Materiali
clear polystyrene
conical bottom clear wells
polystyrene filter
Sterilità
non-sterile
Confezionamento
pkg of 25 ea
Produttore/marchio commerciale
Whatman 7700-2803
Whatman Article No. 28420806 (US reference)
Volume massimo
800 μL
Misura
96 wells
Volume d’esercizio
800 μL
Tipo di binding
non-treated surface
Categorie correlate
Caratteristiche e vantaggi
- No cross talk. Patented integral filter design prevents well-to-well cross contamination.
- Economical to use. Wide range of well volume options ensures efficient use of materials.
- Better control. Choice of filter media allows control of the flow rates and retention characteristics.
- Versatile. A broad range of filtration media is available, including glass fiber, polypropylene, cellulose nitrate, cellulose acetate, nylon, and ion exchange cellulose.
Altre note
Note legali
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
WHA77002803 | 4061826077399 |