Documenti fondamentali
TMODS2185-0008
Nalgene® economy wide-mouth bottle
amber high-density polyethylene, volume 250 mL, case of 72 ea
Sinonimo/i:
amber HDPE bottle, nalgene HDPE bottle
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
Materiali
amber high-density polyethylene
Confezionamento
case of 72 ea
Produttore/marchio commerciale
Nalgene DS2185-0008
Capacità
250 mL
Volume
250 mL
Descrizione generale
Perform short-term general purpose laboratory and field applications with Thermo Scientific Nalgene Wide-Mouth HDPE Economy Bottles with Closure. These light-duty translucent bottles are a lightweight version of the original Nalgene bottle, bulk-packed for ultimate economy. Thinner-walled and more flexible than the Nalgene Lab Quality Bottles, they feature a wide mouth for easy filling and offer excellent chemical resistance to most acids, bases and alcohols.
Note legali
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)