TMO3664-0025
Nalgene® economy graduated cylinder
polypropylene, volume 25 mL, case of 18 ea
Sinonimo/i:
PP graduated cylinder, PP graduated flask
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.43
Prodotti consigliati
Materiali
polypropylene
Confezionamento
case of 18 ea
Produttore/marchio commerciale
Nalgene 3664-0025
Capacità
25 mL
Volume
25 mL
Categorie correlate
Descrizione generale
Dispense liquids easily with simple to read, molded-in graduations that won′t rub off. Affordably priced Thermo Scientific Nalgene Polypropylene Economy Graduated Cylinders have a generous pour spout and exhibit excellent chemical and break resistance.
Note legali
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
TMO3664-0025 | 4061841584971 |