Documenti fondamentali
TMO312006-9125
Nalgene® diagnostic bottle
natural polypropylene copolymer, volume 4 mL, case of 2000 ea
Sinonimo/i:
PPCO bottles, small volume bottle with closure
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
Materiali
natural polypropylene copolymer
Confezionamento
case of 2000 ea
Produttore/marchio commerciale
Nalgene 312006-9125
Capacità
4 mL
Volume
4 mL
Descrizione generale
Sample, store and ship small volume, valuable reagents. Thermo Scientific Nalgene Natural PPCO Diagnostic Bottles with Closure are durable, autoclavable and used in test kits and many small-volume applications. These natural, translucent polypropylene copolymer bottles feature uniform walls resistant to splitting or puncturing. Bottles include linerless polypropylene closures that ensure leakproof† performance without a liner that can wrinkle, cause leaks and contaminate your solutions.
Note legali
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries