Documenti fondamentali
TMO312004-9050
Nalgene® diagnostic bottle
amber high-density polyethylene, volume 15 mL, case of 2000 ea
Sinonimo/i:
PPCO bottles, small volume bottle with closure
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
Materiali
amber high-density polyethylene
Confezionamento
case of 2000 ea
Produttore/marchio commerciale
Nalgene 312004-9050
Capacità
15 mL
Volume
15 mL
Descrizione generale
Sample, store and ship small volume, valuable reagents. Thermo Scientific Nalgene Natural PPCO Diagnostic Bottles with Closure are durable, autoclavable and used in test kits and many small-volume applications. These natural, translucent polypropylene copolymer bottles feature uniform walls resistant to splitting or puncturing. Bottles include linerless polypropylene closures that ensure leakproof† performance without a liner that can wrinkle, cause leaks and contaminate your solutions.
Note legali
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries