TMO267556
Nunc® Edge 2.0 96-well Plate
untreated, polystyrene, sterile, without lid, clear, case of 160 ea
Sinonimo/i:
cell culture plate, cell growth plate
About This Item
Codice UNSPSC:
41122102
NACRES:
NB.15
Materiali
clear
polystyrene
Sterilità
sterile
Confezionamento
case of 160 ea
Produttore/marchio commerciale
Nunc 267556
Categorie correlate
Descrizione generale
Minimize plate evaporation and the risks associated with edge effect by using the uniquely engineered Thermo Scientific Nunc Edge 2.0 Plate. The surrounding bulit-in reservoirs, when filled with sterile water or media, act as evaporation barriers, yielding consistency across the entire 96-well plate. Enabling the use of all 96 wells in a plate allows researchers to plan experiments more efficiently with enhanced productivity and less waste for cell-based assays that require extended incubation.
Note legali
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)