Documenti fondamentali
TMO267427
Nunc® Edge 2.0 96-well Plate
untreated, polystyrene, sterile, with lid, clear, case of 50 ea
Sinonimo/i:
cell culture plate, cell growth plate
About This Item
Codice UNSPSC:
41122102
NACRES:
NB.15
Materiali
clear
polystyrene
Sterilità
sterile
Confezionamento
case of 50 ea
Produttore/marchio commerciale
Nunc 267427
Descrizione generale
Minimize plate evaporation and the risks associated with edge effect by using the uniquely engineered Thermo Scientific Nunc Edge 2.0 Plate. The surrounding bulit-in reservoirs, when filled with sterile water or media, act as evaporation barriers, yielding consistency across the entire 96-well plate. Enabling the use of all 96 wells in a plate allows researchers to plan experiments more efficiently with enhanced productivity and less waste for cell-based assays that require extended incubation.
Note legali
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)