TMO2425-0504
Nalgene® narrow-mouth Right-to-Know wash bottle
low-density polyethylene, bottle volume 500 mL, isopropanol, case of 24 ea
Sinonimo/i:
Nalgene bottle, plastic bottle
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
Prodotti consigliati
Materiali
low-density polyethylene
narrow-mouth
Confezionamento
case of 24 ea
Produttore/marchio commerciale
Nalgene 2425-0504
Volume del flacone
500 mL
Descrizione generale
Meet regulatory labeling requirements for chemical containers with Thermo Scientific Nalgene Narrow-Mouth Right-to-Know LDPE Wash Bottles, which use standard, instantly recognizable symbols and colors. These low density polyethylene bottles are flexible for easy dispensing and translucent for viewing contents.
Note legali
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)