Documenti fondamentali
OH30073978
Ohaus® Pen pH Meter
Model ST10T-A
About This Item
Codice UNSPSC:
41115603
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Ohaus® 30073978
Campo di misura dei TDS
0.0-100.0 mg/L
Batteria
4 × 1.5 V, 200 hr life
Descrizione generale
With durable IP67 waterproof ABS housing, protective sensor cap, and an automatic shutdown feature that preserves battery life, OHAUS pen meters can endure consistent use in rough, wet environments.
Applicazioni
pH, oxidation-reduction potential (ORP), conductivity, dissolved oxygen (DO), salinity, total dissolved solids (TDS), temperature measurement
Caratteristiche e vantaggi
Easily replaceable electrodes, automatic shut off feature
Note legali
Ohaus is a registered trademark of Ohaus Corp.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)