MTCR1040G
MTC™ Bio Fraction CollectorTube Rack
holds 80 x 1.5 /2.0 mL tubes, green, pack of 5 ea
Sinonimo/i:
multi gel processing box, tube holder, tube storage
About This Item
Codice UNSPSC:
41122804
NACRES:
NB.12
Prodotti consigliati
Materiali
green
polypropylene
Caratteristiche
holds 80 x 1.5 /2.0 mL tubes
Confezionamento
pack of 5 ea
Produttore/marchio commerciale
MTC™ Bio R1040-G
Categorie correlate
Descrizione generale
Compact, convenient 80 well racks designed to keep your tubes neatly and efficiently organized. Theses autoclavable polypropylene racks are compatible with conical and self-standing microtubes and all wells have raised, molded numbers for quick identification. The bottom of each well has been recessed to improve conical tube stability without sacrificing the stability of self-standing tubes.
Tube Rack, fraction collector type, 80 x 1.5mL/2mL, orange, pack of 5
Caratteristiche e vantaggi
- 80 well (5 rows of 16) for 1.5 to 2.0mL microtubes
- Rainbow pack available — 5 colors
- Two frosted marking areas, on the bottom and side
- Raised numbers beside each well
- Recessed well bottoms for improved tube stability
Note legali
MTC is a trademark of MTC Bio
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
MTCR1040G | 4065270485878 |
MTCR1040G-5EA | 4065271097094 |