MTCC2051
MTC™ Bio Locktop Style Microtube
capacity 1.5 mL, pack of 500 ea
Sinonimo/i:
centrifuge tube, plastic tube
About This Item
Codice UNSPSC:
41121703
NACRES:
NB.78
Prodotti consigliati
Materiali
clear
polypropylene
round
Sterilità
non-sterile
Caratteristiche
graduation
Confezionamento
pack of 500 ea
Produttore/marchio commerciale
MTC™ Bio C2051
Capacità
1.5 mL
Compatibilità
suitable for PCR
Descrizione generale
MTC™ Bio 0.5 mL microtube come with Locktop style caps. Designed for secure sample storage, the locktop caps ensure a tight seal to prevent any leakage or contamination. These microtubes are ideal for various laboratory applications such as sample storage, PCR, and centrifugation. Their robust construction enables them to withstand high speeds in centrifugation processes without compromising sample integrity. The clear polypropylene material allows for easy visual inspection of the samples. These microtubes are the perfect choice for any lab seeking reliable and convenient sample containment solutions.
Microtube w/ cap, 1.5ml, Locktop style cap, 500/pk
Caratteristiche e vantaggi
- Compatible with standard equipment
- Centrifuge at up to 28,000xg
- RNase, DNase & pyrogen free
- Unsurpassed Clarity
Note legali
MTC is a trademark of MTC Bio
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)